Order Entry
Canada
ContactUsLinkComponent
Eppendorf® ThermoStat Plus Hot/Cold Incubator
Eppendorf® ThermoStat Plus Hot/Cold Incubator
Catalog # CA14400-031
Supplier:  Eppendorf
Eppendorf® ThermoStat Plus Hot/Cold Incubator
Catalog # CA14400-031
Supplier:  Eppendorf
Supplier Number:  022670603

Specifications

  • Description:
    CombiBox Universal block for tubes up to 60mm high (not shown)
  • Cat. no.:
    CA14400-031
  • Supplier No.:
    022670603

Specifications

About this item

Eleven different interchangeable Thermoblocks hold micro tubes, 15 mL or 50 mL conical tubes, or microplates. The CombiBox Universal Block holds different sized vials and beakers.

  • Store and recall up to 4 independent programs with set temperatures and times
  • New, exchangeable thermoblocks for slides and microarrays available
  • RS-232 interface provides control and documentation in accordance with GLP
  • 2-year warranty