Eppendorf 5427R Microcentrifuges
Supplier: Eppendorf
Certificates
About this item
Labs demanding high sample throughput will benefit from the 5427R, featuring a double-row 48-place rotor to increase efficiency and improve running time.
- 48 x 1.5/2.0 mL capacity rotor available
- Eppendorf QuickLock lids come standard with all aerosol-tight rotors
- Automatic rotor recognition and imbalance detection for maximum safety
- Maintains constant 4°C at maximum speed
- FastTemp program is optimized for super-fast 11-minute pre-cooling and maximum temperature accuracy inside the rotor
The 5427R provides refrigeration with innovative compressor technology that continuously cools even after the run, so your samples stay safe.
Customers Who Bought This Also Bought