Orders LinkContactUsLinkComponent
MBP Flip-Strip™ Racks
MBP Flip-Strip™ Racks
MBP Flip-Strip™ Racks
Certificates
MBP Flip-Strip™ Racks
MBP Flip-Strip™ Racks
1 Options Available Below
Product Details & Documents
This reversible rack is ideal for any researcher utilizing strip tubes for PCR.
  • Made from recycled plastic
  • Stable, low-profile design with an alphanumeric grid for easy tube identification
  • FlipStrip racks come complete with a snug fitting lid
  • Lid protects samples and allows racks to be stacked
  • Several colors available
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Red, Blue, Yellow and Green
    Discontinued
    Supplier Number: -
  • Green
    Discontinued
    Supplier Number: -
  • Blue
    Discontinued
    Supplier Number: -
  • Purple
    Discontinued
    Supplier Number: -
  • Fluorescent Orange
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $80.50
    $80.50
    Log in for availability
    Case of 12
    Case of 12 - $1,763.33
    $1,763.33
    New Customer Promotional Price
    Each
       $80.50
    Case of 12
       $1,763.33
    Supplier Number: 8678
  • Discontinued
    Supplier Number: -