MBP Locking-Cap Microcentrifuge Tubes, Graduated
Catalog # 490016-243
Supplier: Thermo Scientific
Specifications
- Color:Assorted
- Description:0.6 ml microcentrifuge tubes with locking cap
- Cat. no.:490016-243
- Supplier No.:L-502-GRD-MIXD
Specifications
About this item
A superior lock for safe and convenient centrifuging, boiling, freezing, incubation or storage of samples.
- Smooth surfaces reduce the possibility of ripped or punctured gloves
- Manufactured of ultra clear, autoclavable polypropylene
Locking microcentrifuge tubes offer the same sample security as found with screw cap tubes yet maintain the same ease of use as a snap cap. Unique design allows visual confirmation that the tube is locked. The pierceable cap snaps closed and the lock slips easily into place with a simple one hand operation. The larger, rounder cap design proves less stressful to the thumb.