Eppendorf Reagent Reservoir
Catalog #: CA53513-048
Supplier: Eppendorf
Low Manufacturing Impact
Specifications
- Capacity:50 - 60 ml
- Description:Reagent Reservoir
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Supplier No.:022265806
Specifications
About this item
Convenient autoclavable reservoir.
Easy-to-use loading of multichannel pipettes.
Customers Who Bought This Also Bought