Teknova Complete Minimal Dropout Plates Glucose with Four Dropouts
Catalog # CA100217-580
Supplier: Teknova
CAS Number:
Specifications
- Description:CM Glucose Plates minus Histidine, Leucine, Tryptophan, Uracil
- Cat. no.:CA100217-580
- Plate Size:100 mm
- Supplier No.:C3700
Specifications
About this item
Plates are shelf-life dated and guaranteed for 4 weeks from the date of shipment
All plates must be stored at 4°C and in the absence of continuous exposure to light. With proper storage, most antibiotic plates are useable for up to 2 months. All plates are sold in sleeves of 20 unless noted otherwise. Complete Minimal Dropout Plates (CM), also known as Synthictic Dropout Plates (SD).