HyperSep Glass Block Manifolds, Thermo Scientific
Supplier: NATIONAL SCIENTIFIC MX
Certificates
About this item
Provides multiple processing of SPE or SLE cartridges to improve throughput and reproducibility.
Delivery: Includes glass block manifold lid, cover gasket, vacuum gauge, valve assembly, tips (16 or 24), adjustable collection rack, bulkhead luer fittings, plugs (16 or 24) and manifold safety tray.
Frequently Bought Together