Nalgene® Economy Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Economical, single-use, break-resistant bottles feature thinner walls and are more flexible than original Nalgene® bottles.
- Excellent chemical resistance to most acids, bases, and alcohols
- Guaranteed leakproof
- Available in natural or amber
- Narrow mouths are ideal for pouring
Ideal for short-term, general-purpose laboratory and field applications, such as collecting samples, or for storing working solutions like buffers, bench reagents, and diluted chemical aliquots. Natural bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Certifications: Amber bottles comply with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Delivery: Supplied with polypropylene screw closures.
Caution: Not recommended for hazardous and critical material containment.
Specifications
- Catalog No:
- 16172-104
- 16172-110
- 16172-126
- 16172-128
- 16172-130
- 16172-124
- 16172-122
- Environmentally Preferable:
- Low Manufacturing Impact
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Product Transparency and Disclosure
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ACT Label
- ACT Label
- ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
- ACT Label