ContactUsLinkComponent
Accucore™ Phenyl-X Columns, Thermo Scientific
Accucore™ Phenyl-X Columns, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
undefined
Accucore™ Phenyl-X Columns, Thermo Scientific
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 10039-016
  • 10039-018
  • 10039-020
  • 10039-024
  • 10039-026
  • 10039-030
  • 10039-032
  • 10039-034
  • Description:
  • 10039-016
  • 10039-018
  • 10039-020
  • 10039-024
  • 10039-026
  • 10039-030
  • 10039-032
  • 10039-034
  • Carbon load:
  • 6%
  • 6%
  • 6%
  • 6%
  • 6%
  • 6%
  • 6%
  • 6%
  • pH range:
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • Surface area:
  • 130 m²/g
  • 130 m²/g
  • 130 m²/g
  • 130 m²/g
  • 130 m²/g
  • 130 m²/g
  • 130 m²/g
  • 130 m²/g