ContactUsLinkComponent
Syncronis™ Silica HPLC Columns, Thermo Scientific
Syncronis™ Silica HPLC Columns, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
undefined
Syncronis™ Silica HPLC Columns, Thermo Scientific
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 10044-052
  • 10044-012
  • 10044-056
  • 10044-026
  • 10044-048
  • 10044-020
  • 10044-042
  • Description:
  • 10044-052
  • 10044-012
  • 10044-056
  • 10044-026
  • 10044-048
  • 10044-020
  • 10044-042
  • pH range:
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • Surface area:
  • 320 m²/g
  • 320 m²/g
  • 320 m²/g
  • 320 m²/g
  • 320 m²/g
  • 320 m²/g
  • 320 m²/g