ContactUsLinkComponent
Syncronis™ Phenyl HPLC Columns, Thermo Scientific
undefined
Syncronis™ Phenyl HPLC Columns, Thermo Scientific
  • Catalog No:
  • 10044-234
  • 10044-278
  • 10044-286
  • Description:
  • 10044-234
  • 10044-278
  • 10044-286
  • Carbon load:
  • 11%
  • 11%
  • 11%
  • pH range:
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • Surface area:
  • 320 m²/g
  • 320 m²/g
  • 320 m²/g
Frequently Bought Together