GFM Pro Electronic Flowmeter, Thermo Scientific
Catalog #: 10057-406
Supplier: Thermo Scientific
Specifications
- Description:GFM Pro Flowmeter
- Cat. no.:10057-406
- Supplier No.:66002-010
Specifications
About this item
The Thermo Scientific GFM Pro Flowmeter is specifically designed for use with gas chromatography instruments.
- Portable unit and can be hand-held
- Has optional stand for bench-top convenience
This versatile product is an electronic device capable of measuring volumetric flow for all types of gases. Real-time measurements can be made for various types of flow paths including continually changing gas types.
Customers Who Bought This Also Bought