Acclaim™ PepMap™ 100 C18 Columns
Supplier: Thermo Scientific
Certificates
About this item
This is the standard column for high-resolution, high-sensitivity protein identification for proteomics and biomarker discovery and verification.
The Thermo Scientific™ Acclaim PepMap column is ideal for high-resolution peptide mapping of tryptic, natural, and synthetic peptides and LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology.
Specifications
- Catalog No:
- 10040-654
- 10040-652
- 10818-734
- 10044-940
- 10818-732
- 10040-694
- 10818-574
- 10818-572
- 10040-658
- 10818-576
- 75802-920
- 10040-670
- 10040-686
- 10040-640
- 10040-646
- 10040-644
- 10040-690
- 10818-736
- 10040-688
- Description:
- NanoViper Trap Column
- 10040-652
- 10818-734
- 10044-940
- 10818-732
- 10040-694
- 10818-574
- 10818-572
- 10040-658
- 10818-576
- 75802-920
- 10040-670
- 10040-686
- Trap Column
- Trap Column
- Trap Column
- 10040-690
- Trap column
- 10040-688
- Carbon load:
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- 17400 psi
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- Storage:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
Specifications
Frequently Bought Together