Order Entry
Canada
ContactUsLinkComponent
Labnet ENDURO™ MiniMix, 230 V
Labnet ENDURO™ MiniMix, 230 V
Catalog # 490009-312
   Low Manufacturing Impact
undefined
Labnet ENDURO™ MiniMix, 230 V
Catalog # 490009-312
Supplier Number:  S0600
   Low Manufacturing Impact

Specifications

  • Temperature Range:
    4...65 °C
  • Shaking motion:
    3-D
  • Description:
    Enduro MiniMix
  • Dimensions:
    24 x 17.15 x 15 cm (9.75 x 6.75 x 6")
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Electrical:
    230V~, 50 Hz or 120V~, 60 Hz
  • Max. load:
    0.8 kg (1.75 lbs.)
  • Pitch angle:
    Fixed 5°
  • Weight:
    0.88 kg (1.9 lbs.)
  • Cat. no.:
    490009-312
  • Maximum Load:
    0.8 kg (1.75 lbs.)
  • Speed:
    18 rpm
  • Supplier No.:
    S0600

Specifications

About this item

Labnet's Enduro MiniMix combines the motions of an orbital shaker and a rocker to produce three dimensional action. The benefit of this type of motion is that it is both gentle and thorough.

  • 230 V model
  • Designed for processing blots and staining gels
  • Optimal fixed speed and tilt
  • Three dimensional motion
  • Enduro Blot boxes are perfect for protein gels

The pitch and speed of the Enduro MiniMix are fixed to provide the optimal conditions for processing western blots and staining gels. The tilt angle is large enough to provide thorough distribution of solutions, yet small enough to keep the center of the blot from drying out. Minimal amounts of solution can be used, conserving valuable probes and antibodies.

The mixer is supplied with a non-slip rubber mat to help keep boxes in place on the platform. The housing and platform are easy to clean and decontaminate when needed. For temperature controlled incubations, the Enduro MiniMix may be used in a cold room or an incubator. The unit is lightweight and compact and easily moved where needed.

Labnet Blot Boxes are available in two sizes. The small boxes have a capacity for 3-5 mL of solution and are perfect for mini protein gels, such as those used in the Enduro Mini PAGE System. The large boxes will hold 6-10 mL and are sized for larger mini protein gels. The boxes are acrylic and feature a hinged lid with a positive catch. The boxes are supplied in packages of 10.

Caution: 230 V model.