GlacierBrand™ Ice Buckets with Lids, Diversified Biotech
Excellent insulation is made of triple density expanded polystyrene
Preserves ice 30% longer. Thick pan walls will not sweat or leave puddles of condensation on the bench. Robust handles make buckets easy to carry. Use the supplied lid to maximize cooling. Solidly constructed to be unbreakable under normal laboratory conditions. Resistant to mild chemicals as well as ice-alcohol slurry, dry ice, and liquid nitrogen.
Frequently Bought Together