SP Bel-Art Spinwedge Magnetic Stirring Bars, Bel-Art Products, a part of SP
Spinwedge® magnetic stirring bars provide strong turbulence at fairly low speeds and are well suited for churning sediment or dissolving salts.
- Wedge shape is designed to plow up sediment for thorough dispersion
- Sharply angled broad face provides strong mixing action
- PTFE low-friction coating
- Durable and inert
- Autoclave safe
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Frequently Bought Together