Nylon Cloth Labels, GA International
Supplier: Avantor
Certificates
About this item
These labels are made from matte white polyamide coated nylon cloth material with permanent acrylic adhesive and compatible to thermal transfer printers.
- Recommended for indoor use
- All-in-one cartridge includes label supply and R4300 Series ribbon
Ideal for performance and general purpose lab labeling, the permanent adhesive and thicker construction makes this material durable, yet easy to handle. Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
Frequently Bought Together