Orders LinkContactUsLinkComponent
Labels, Brady
Supplier:   Avantor
Labels, Brady
Supplier:  Avantor
Certificates
Labels, Brady
5 Options Available Below
Product Details & Documents
Labels for the TLS 2200 and TLS-PC Link Thermal Transfer Portable Printers.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Please Note: This material is UL, CSA and AGA approved with its respective ribbon. See the 'Recommended Ribbon Series' for the appropriate ribbon to use.
Product Family Options
Product Information
  • Material
    Dimensions
    Packaging
    Availability
    Price
  • CA80090-086
    Portable thermal labels
    Portable thermal labels
    Nylon Cloth
    12.7×38.1 mm (0.5×1.5")
    500 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - 103,40 $
    103,40 $
    Each
       103,40 $
  • CA80090-092
    White nylon cloth labels
    White nylon cloth labels
    Nylon Cloth
    48.26×25.4 mm (1.9×1")
    250 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - 132,30 $
    132,30 $
    Each
       132,30 $
  • CA80090-126
    Portable thermal labels
    Portable thermal labels
    High Performance Polyester
    19×22 mm (0.75×0.9")
    250 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - 52,02 $
    52,02 $
    Each
       52,02 $
  • CA80090-136
    Self-laminating labels
    Self-laminating labels
    Self-laminating Polyester
    45.72×12.7 mm (1.8×0.5")
    500 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - 117,85 $
    117,85 $
    Each
       117,85 $
  • CA80090-140
    Portable thermal labels
    Portable thermal labels
    Self-laminating Polyester
    2.200×0.5" (55.880×12.7 mm)
    100 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - 32,01 $
    32,01 $
    Each
       32,01 $