SP Bel-Art Mono-Block™ 12-Well Reservoir, Bel-Art Products, a part of SP
Catalog # 490005-652
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Mono-block 12 Well Reservoir
- Cat. no.:490005-652
- Supplier No.:F37880-1200
Specifications
About this item
Use this reservoir with 8- or 12-channel pipettors for plate washing or feeding.
- Stable base
- V-shaped bottom to improve concentration
- Reduced waste from well design
Each separate well holds 5 mL and has a v-shaped bottom to concentrate fluids and reduce waste. A wide base is stable for bench top use or comfortable to hold in the hand. Mono-Block is a trademark of Bel-Art Scienceware.