Labnet Excel™ Electronic Pipettes, Labnet International
: Avantor
The Excel™ electronic pipettes are multifunctional units with operational modes for standard and reverse pipetting as well as multiple and sequential pipetting, mixing of samples and serial dilutions.
- Exceptional accuracy and precision
- Simple, precise volume setting
- Fully motorized piston drive
- Multifunctional with six operating modes
- Virtually eliminates pipetting force
- User friendly operator interface
- Rechargeable Lithium-ion battery
- Automatic calibration using external weight
All functions are easily programmed with one hand on the well positioned display and keypad. A fully motorized piston drive and auto-calibration guarantee excellent accuracy and reproducibility. A removable, rechargeable Lithium-ion battery ensures many hours of performance between charges.
Frequently Bought Together