BrightMax™ White Sealing Film for Luminescence and Microscopy, Excel Scientific
Catalog # 490007-384
Supplier: Excel Scientific
Specifications
- Optical property:Light blocking
- Sterility:Non sterile
- Description:BrightMax™ White Sealing Film
- Thickness:203 µm
- Cat. no.:490007-384
- Supplier No.:WT-50
Specifications
About this item
These sealing films are 203 m thick white vinyl films with 37m acrylic adhesive for use with white microplates. The sealing films may be applied to either the top or bottom of the plate.
- White light-reflecting sealing films for use with white microplates
- May be applied to top or bottom of plate
- Maximize intensity and signal-to-noise ratios in luminescence assays
- Additional applications for mounting and storage of filters for microscopy, for example in ELISPOT assays
A split backing aids in applying the sealing film properly. The light-reflective properties of BrightMax plate sealing films on clear-bottom plates maximize sensitivity of bioluminescence and chemiluminescence assays in bottom-reading luminometers. White Films also have application as a backing matrix for microscopy of punchout samples from filter plates, for example in ELISPOT assays. Recommended for temperatures from -40°C to 80°C. Dimensions: 76.2 x 116 mm without end tabs.