EZ-Pierce™ Sealing Films, Excel Scientific
Supplier: Excel Scientific
Certificates
About this item
Easily pierceable, 70 m thick, polyethylene with acrylic adhesive, EZ-Pierce sealing films allow direct sample recovery with single or multichannel pipettors and robotic probes.
- Easily pierceable with pipet tips or robotic probes
- Inert, chemical resistant
- Functional temperature range -40°C to 90°C
- Printed alphanumeric template version simplifies well location
They are available with or without a printed 96-well alphanumeric template for easy well location. The printed EZ-Pierce sealing films also have color-coding dots and white marking areas on each end tab for plate identification. Dimensions are 82.6 x 142.9 mm for 96-well plates.
Frequently Bought Together