Specifications
- Description:AccuSeal Semi-Automated Microplate Sealer
- Weight:12.2 kg (27 lbs.)
- Sealing time:0.5 to 10 seconds
- Temperature set range:100 - 190 °C
- Cat. no.:490009-278
- L×W×H:32.4 x 17 x 36.2 cm (12.75 x 6.75 x 14.25")
- Supplier No.:PS1000
Specifications
About this item
Labnet's AccuSeal Semi Automated Plate Sealer is ideal for the low to medium throughput Laboratory that requires uniform and consistent sealing of microplates. Offering complete versatility, the AccuSeal will accept a full range of plates for PCR, assay or storage applications, and can be used for standard height to deep well plates. Operation is easy, sealing parameters are set and displayed via the user-friendly control panel, and the sealing operation is automated to ensure consistent results. Controls and Operation - The AccuSeal's Control System features an intuitive layout, and allows optimal settings for all types of plates and sealing material. Press the MODE button to choose Sealing Time, or Sealing Temperature, and use the convenient turn knob to change settings. Temperature can be precisely set in increments of 1.0°C, and time in increments of 0.1 seconds. The AccuSeal also features a No Heat setting for pressure sealing applications.
- Designed for consistent heat sealing
- Works with PCR, assay, and deep well plates
- Accepts shallow and high profile plates
- Adjustable sealing temperature and timing
- Digital control panel
The sealing procedure is simple. Place a microplate in the open drawer, place a heat seal on top of the plate, and press the SEAL button. The motorized drawer will automatically close, the heated sealing plate will press firmly down on the seal and plate for the precisely set time, and then the drawer will open to present the sealed plate. Labnet's AccuSeal can accept a variety of plate types and heights. An aluminum adapter is used for PCR and standard assay plates. The motorized heated platen adjusts automatically for different height plates, and provides uniform pressure to ensure a quality seal. The platen incorporates a special non-stick coating, but in the event that cleaning is necessary, an access door can be opened on the side of the instrument.
The AccuSeal has been designed with safety in mind. The internal heating elements are situated inside the instrument casing, and an internal safety door prevents access to the heated area. In addition, the motorized drawer features redundant safety switches - if a hand or object obstructs the closing drawer, the drawer motor will reverse. This feature prevents injury to a user as well as to the instrument.
Temperature Accuracy: ±1.0°C. Temperature Uniformity: ± 1.0°C. Compatible Plate Materials: PP (Polypropylene), PS (Polystyrene), PE (Polyethylene). Compatible Plate Types: Standard Assay Plates, Deep-Well Storage plates, PCR Plates (Non-Skirted, Semi-Skirted, Full-Skirted). Maximum Plate Height: 45 mm. Compatible Sealing Film Types: Foil Polypropylene laminate, Clear Polyester-polypropylene laminate, Clear polymer, Thin clear polymer, Foil-laminate, Foil.