Shelton Scientific IBI® Horizontal Gel Systems: High Resolution and High Throughput Design, 20×25 cm
Catalog # 490007-706
Supplier: IBI Scientific
Specifications
- Description:High Resolution Horizontal Gel System
- Cat. no.:490007-706
- Supplier No.:IB56000
Specifications
About this item
This 20 x 25 cm Horizontal Gel Systme is our largest high-resolution unit and is ideally suited for inversions and genomic DNA separations requiring buffer recirculation. All models include buffer chamber, safety cover, power cords, casting system with tray, two combs, leveling bubble, and manual.
- Buffer chamber
- Safety cover and power cords
- Casting system with tray
- Two 2.0 mm thick 20-tooth combs
- Leveling bubble and manual