Ward's® Single Probes Light Sensor
Catalog #: 470014-820
Supplier: Wards
Specifications
- Description:Ward's Single Probes Light Sensor
- Cat. no.:470014-820
- Supplier No.:WSP-104-500
Specifications
About this item
Measure the intensity of light in the darkest of classrooms to brightest school yards with the Ward's Single Probes Light Sensor.
- Measure the intensity of light
- Collect and record data
- Free activity guide
It's easier than ever for students to collect and record light data for experimentation and analysis. Sensor includes free activity guide with activities designed for use with each probe, available digitally online after purchase. Resolution: 0-1000 lx 1 lx, 0-6000 lx 1 lx, and 0-150,000 lx 22 lx. Sample rate, 10,000. Requires Ward's Single Probes Base Unit, available separately.
Caution: Measure the intensity of light
Collect and record data
Free activity guide
Customers Who Bought This Also Bought