Orders LinkContactUsLinkComponent
VWR® Microprobe Set
152 mm
  89259-848
VWR® Microprobe Set
  89259-848
VWR® Microprobe Set
VWR® Microprobe Set
152 mm
$244.34
Each Pack of 12
This VWR® Microprobe Set is for micromanipulation in the lab.
  • Stainless steel construction
  • Needle-sharp tips
  • Variety of microprobes available
Each microprobe is precision-ground stainless steel that has been tempered for quality control and experimental requirements. Tips are needle-sharp and have been hand-honed. Set includes six different microprobes.
  • Length:
    152 mm