Orders LinkContactUsLinkComponent
VWR® Cushing Forceps
Stainless steel, 178 mm (7")
Catalog # 89259-952
Supplier:   VWR International
VWR® Cushing Forceps
Catalog # 89259-952
Supplier:  VWR International
VWR® Cushing Forceps
VWR® Cushing Forceps
Stainless steel, 178 mm (7")
Price
$70.10
New Customer Promotional Price
Log in for availability
Each Pack of 12
Product Details & Documents
VWR® Cushing Forceps are stainless steel with straight, serrated tips.
  • Stainless steel construction
  • Straight, serrated tips
They feature a corrugated handle and a guide pin.
Specifications
  • Length:
    178 mm (7")
  • Material:
    Stainless steel