Semi-Micro Balances, METTLER TOLEDO®
Supplier: Mettler Toledo
Certificates
About this item
These MS Semi-Micro Balances offer strong performance and increased productivity thanks to a fast settling time and consistently reliable results
- Strong performance
- Increased productivity
- Fast settling time
- Reliable results
- ErgoDoors
- Built-in date and time functions
The Swiss high-precision weighing cell HRT guarantees constant accuracy over the entire weighing range. Clever ErgoDoors make the weighing-in of samples as easy and convenient as possible. The built-in date and time function enables GLP/GMP compliant print outs, ensuring full control and traceability for your research.
Related Information
F&BFOCUS V4 Issue 2
Product Literature
Mettler-Toledo Balance or Analytical Instrument Mettler-Toledo Balance or Analytical Instrument_CA
Promotion Material
NewClassic MS Brochure
Datasheet
Customers Who Bought This Also Bought