IBI Plant Isolate DNA Extraction Kit, IBI Scientific
Supplier: IBI Scientific
Certificates
About this item
IBI Plant Isolate Total DNA Extraction Reagent System provides a quick and easy 3 step CTAB and chloroform based method to isolate Total DNA (including genomic, mitochondrial, and chloroplast DNA) from a variety of plant species (including algae and cyanobacteria)
This unique reagent is able to lyse most common plant samples as well as plant samples with a high polysaccharide content. The extracted DNA is suitable for routine PCR Screening, Real-Time PCR, Southern Blotting, Mapping, and RFLP. Phenol extraction is not required and the entire procedure can be completed within 50 minutes.
Ordering information: Kit contains IBI Plant Isolate and Rnase A (50 mg/mL).
Specifications
- Refine target molecule:Total DNA
- Sample type:Plant samples
- Target molecule:DNA
- Sample size:Up to 1 g of Fresh Plant Tissue; Up to 0.5 g of Dry Plant Tissue
- Operation time:50 min
Specifications
Frequently Bought Together