Soil Standard, Oil & Grease in Soil, ERA (Waters)
Catalog # CA11022-780
Supplier: ERA
CAS Number:
Specifications
- Description:Certified Reference Material
- Size:50 g
- Cat. no.:CA11022-780
- Supplier No.:549
- Parameters:Oil and Grease in Soil
Specifications
About this item
Use with gravimetric method 9071B or infrared spectrometric analysis of soil.
The analytes are in the specified NELAC range of 300-3000 mg/kg.
Methods: EPA 9070A, EPA 9071 B, EPA 1664A, EPA 1664B, EPA 413.1, EPA 413.2, EPA 418.1, EPA 8440, EPA 9070, EPA 9071A, EPA 9071B, SM 5220 B, SM 5220 C, SM 5220 D, SM 5520 D, SM 5520 E, SM 5520 F
Ordering information: One 50 g screw-cap bottle of soil is ready to be analyzed