Lava Soap Bar, Kranz
Catalog #: 11217-202
Supplier: Kranz
Specifications
- Description:Lava soap bar, 4 oz.
- Cat. no.:11217-202
- Supplier No.:19218
Specifications
About this item
The volcanic pumice contained in the soap possesses abrasive qualities that give it the scrubbing power to clean dirt, grease, oil, ink, tar, paint, caulk and adhesives from hands
Lava hand soap is ideal for use in automobile repair, warehouses, manufacturing facilities and any occupation where dirt, grease and grime are present.
Customers Who Bought This Also Bought