ContactUsLinkComponent
SP Bel-Art Glove Box Port Covers, 8" ext.Ø., Bel-Art Products, a part of SP
SP Bel-Art Glove Box Port Covers, 8" ext.Ø., Bel-Art Products, a part of SP
  54848-128
undefined
SP Bel-Art Glove Box Port Covers, 8" ext.Ø., Bel-Art Products, a part of SP
  54848-128
 :  H50025-0008
  • Description:
    Covers for 8" ports
  • Cat. no.:
    54848-128
  • Supplier No.:
    T50025-0008
Use these glove box port covers to seal off any glove ports to which gloves are not attached.
  • Single-piece design
  • Neoprene gasket provides a snug fit
  • Easy to apply port covers
Covers are one-piece construction with a neoprene gasket for a snug fit and fit glove ports with a 20.3 cm (8") O.D. Simply press the covers into the port opening until the flange on the cover meets the glove port.