Hardness (Ca and Total Sequential) HR Reagent Set, Digital Titrator, Hach
Catalog # CA11027-144
Supplier: Hach
CAS Number:
Specifications
- Chemical method:EDTA
- Cat. no.:CA11027-144
- Range:100 - 4000 mg/L CaCO3
- Supplier No.:2448700
- No. of Tests:100
Specifications
About this item
For sequential determination of Calcium and Total Hardness by EDTA Digital Titrator titration.
- Accurate Results; when and where you need them
- Fast and Flexible
- Versatile
- Economical
Hach method 8329. Range: 100-4000 mg/L CaCO3. Set contains Buffer Solution (Hardness 1), CalVer® 2 Calcium Indicator Powder Pillows, ManVer® 2 Hardness Indicator Powder Pillows, Potassium Hydroxide Solution (8 N), Sulfuric Acid (5.25 N) and 0.800 M EDTA Digital Titrator Cartridge. Approximately 40-100 tests.
Ordering information: Hazardous Items; items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.