Order Entry
Canada
ContactUsLinkComponent
Chloride Reagent Set, Mercuric Nitrate Digital Titrator, Hach
Chloride Reagent Set, Mercuric Nitrate Digital Titrator, Hach
Catalog # CA11024-834
Supplier:  Hach
CAS Number:  
undefined
Chloride Reagent Set, Mercuric Nitrate Digital Titrator, Hach
Catalog # CA11024-834
Supplier:  Hach
Supplier Number:  2272600
CAS Number:  

Specifications

  • Chemical method:
    Mercury nitrate
  • Cat. no.:
    CA11024-834
  • Range:
    10 - 8000 mg/L Cl-
  • Supplier No.:
    2272600
  • No. of Tests:
    100

Specifications

About this item

For determination of chloride by mercuric nitrate Digital Titrator titration

Note: Contains mercury. Dispose of according to local, state and federal regulations.

Hach Method 8206. Ranges: 10-160 and 100-8000 mg/l Cl-. Reagent Set includes: Diphenylcarbazone Reagent Powder Pillows, Mercuric Nitrate Digital Titrator Cartridges, 0.2256 N and 2.256 N. Approximately 40-100 tests.

Ordering information: Hazardous Items; items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.