ContactUsLinkComponent
Chlorine (Total) Reagent Set for CN-21P Test Kit, Hach
Chlorine (Total) Reagent Set for CN-21P Test Kit, Hach
Catalog # CA11014-668
Supplier:  Hach
CAS Number:  
undefined
Chlorine (Total) Reagent Set for CN-21P Test Kit, Hach
Catalog # CA11014-668
Supplier:  Hach
Supplier Number:  2437700
CAS Number:  
Specifications
  • Chemical method:
    Iodometric
  • Description:
    Chlorine (Total) Reagent Set for CN-21P Test Kit
  • Cat. no.:
    CA11014-668
  • Supplier No.:
    2437700
  • No. of Tests:
    100
Specifications
About this item
For determination of Chlorine (Total) by thiosulfate drop-count titration
Replacement reagent set for CN-21P Test Kit. Set includes Sulfamic Acid Powder Pillows, Potassium Iodide Powder Pillows and Sodium Thiosulfate Standard Solution (0.0246 N).