ContactUsLinkComponent
SP Bel-Art Bulb Pipettor, Bel-Art Products, a part of SP
SP Bel-Art Bulb Pipettor, Bel-Art Products, a part of SP
Catalog # 70690-152
undefined
SP Bel-Art Bulb Pipettor, Bel-Art Products, a part of SP
Catalog # 70690-152
Supplier Number:  H37888-0000
Specifications
  • Description:
    Bulb Pipettor
  • Cat. no.:
    70690-152
  • Supplier No.:
    378880000
Specifications
About this item
A tapered, silicone rubber seal provides an airtight fit on all size pipettes.
  • Tapered design
  • Silicone rubber seal
  • Autoclavable
An excellent aspirator for use with Westergren or Wintrobe sedimentation tubes. The bulb can be opened, cleaned, and steam autoclaved at 121 °C (250 °F).
Delivery: Supplied complete with an elastic cord for dedicating the pipettor to a specific reagent bottle.