Order Entry
Canada
ContactUsLinkComponent
 
Tweezers, Straight Flat Strong, Excelta Corp®
  89411-790
 :  Excelta
undefined
Tweezers, Straight Flat Strong, Excelta Corp®
  89411-790
 :  Excelta
 :  8-SA-PI

 

  • Description:
    Tweezers, lead straightening, 2 star quality
  • Material:
    Stainless steel 316L, anti-magnetic
  • Curvature:
    Straight
  • Cat. no.:
    89411-790
  • Overall Length:
    106.25 mm (4.25")
  • Supplier No.:
    8-SA-PI

 

 

Designed for straightening leads and wires.

  • Large flat tips for straightening leads/flashing
  • Stainless steel, corrosion resistant

The 8-SA-PI two star straight broad body flat sharp point tweezer is 4.25" long is ideal for straightening small wires or leads. It is made of anti-magnetic stainless steel.