If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Arkansas Sharpening Stone Hard (Fine)
- Cat. no.:100502-466
- Supplier No.:62082-10
These stones are natural with close density oil-tones, and excellent qualities that make them the stone unequaled for sharpening tools such as gravers, knives, etc.
These sharpening stones comes in a cedar wood box and they are available in two types: translucent (extra fine); and hard (fine).
Translucent Stone Size: 102 x 51 x 12.7 mm (4 x 2 x 1/2")
Hard Size: 152 x 51 x 12.7 mm (6 x 2 x 1/2")