If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Conductive Gold-Paste Extender
- Cat. no.:102090-970
- Supplier No.:12643
Specifications
About this item
This EMS Conductive Gold-Paste is a one part adhesive.
- Fast drying
- Dries at room temperature
- Useful for testing and temporary work
Maximum service temperature is 65°C. This adhesive is not for permanent use; useful for testing and temporary work where a high signal is required from the adhesive. Gold content is ~75%, including sphere sizes 2 µm, and flake size 10 µm, in organic binders and a solvent. Keep refrigerated for good shelf life. Sheet resistance is 0.02 to 0.05 ohm-cm at 1 mil thickness.