ContactUsLinkComponent
Backscattered Electron Detector Calibration Standard, Electron Microscopy Sciences
Backscattered Electron Detector Calibration Standard, Electron Microscopy Sciences
Catalog # 102099-006
CAS Number:  
undefined
Backscattered Electron Detector Calibration Standard, Electron Microscopy Sciences
Catalog # 102099-006
Supplier Number:  80070-BED
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Backscattered Electron Detector Calibration Standard
  • Cat. no.:
    102099-006
  • Supplier No.:
    80070-BED
Specifications
About this item
This MAC standard checks detector and SEM performance.
It contains standards to check one atomic number resolution, Faraday cup to set probe current and duplex brass to check 0.1 atomic number resolution.
Certified standards for system calibration. Carbon block measures 30×5 mm.