Fuchs-Resenthal Count Chamber, Electron Microscopy Sciences
Product Details & Documents
Designed for counting leukocytes, eosinophils and cells in cerebrospinal fluid.
Grid pattern is etched directly onto the glass surface. The chamber is 0.2 mm deep. Chamber set includes: one chamber and 2 octagonal cover glasses.
Specifications
- Description:Fuchs Resenthal Counting Chamber (3720)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)