Orders LinkContactUsLinkComponent
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini-Cooling Chambers, Electron Microscopy Sciences
Certificates
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini-Cooling Chambers, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for ice pans and ice buckets for use when only a small number of microtubes are required.
Each Mini-Chamber will fit in our 1L Glacierbrand™ Ice Pan (6.13 x 6.13 x 3.61"). Minimize Ice - Less Ice is needed to cool these chambers. Less ice means less mess. Mini-Chambers are anodized aluminum and utilize our "Surround-A-Tube" cooling technology.
Product Family Options
Product Information
  • Availability
    Price
  • 102096-362
    Mini Cooling Chamber, 6 Holes
    Mini Cooling Chamber, 6 Holes
    Discontinued
    Supplier Number: 68600-10
  • 102096-364
    Mini Cooling Chamber , 15 Holes
    Mini Cooling Chamber , 15 Holes
    Log in for availability
    Log in for availability
    Each
    Each - $721.81
    $721.81
    Each
       $721.81
    Supplier Number: 68600-20
  • 102096-366
    Mini Cooling Chamber 30 Holes
    Mini Cooling Chamber 30 Holes
    Discontinued
    Supplier Number: 68600-30
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.