ContactUsLinkComponent
Aqua-Hold Pap Pen, Electron Microscopy Sciences
Aqua-Hold Pap Pen, Electron Microscopy Sciences
Catalog # 102091-216
CAS Number:  
undefined
Aqua-Hold Pap Pen, Electron Microscopy Sciences
Catalog # 102091-216
Supplier Number:  71311
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Aqua-Hold Pap Pen
  • Cat. no.:
    102091-216
  • Supplier No.:
    71311
Specifications
About this item
The Aqua-Hold Pap Pen draws a liquid repellent barrier around a section of tissue. Used like a paint pen, you just draw a well on a slide. The barrier creates the proper surface tension to hold aqueous solutions such as costly antibody and other valuable specimens.
  • After application, the barrier material is insoluble in alcohol and acetone but can be removed after staining with xylene
  • Each pen holds 13 mL of long-lasting material, making hundreds of wells
  • Red tint distinguishes the well drawing on the slide to easily view