Orders LinkContactUsLinkComponent
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Certificates
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This complete selection of Wheaton Micro Tissue Grinders is conveniently packed in a lightweight protective carrying case. The 0.5 ml micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
  • Complete kit includes 7 micro tissue grinders
  • Made from Wheaton 33 low extractable borosilicate glass
  • Replacement parts available separately
  • Autoclavable
This particular tissue grinder mortar can be safely joined with another threaded glass component if needed, using the Wheaton Connection®. The Kit is packed in high-density polyethylene case with foam inserts.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 102091-106
    Complete Kit
    Complete Kit
    Log in for availability
    Log in for availability
    Each
    Each - $2,019.97
    $2,019.97
    Each
       $2,019.97
    Supplier Number: 64800
  • 102095-700
    Tissue Grinder
    Tissue Grinder
    0.2 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $423.99
    $423.99
    Log in for availability
    Pack of 2
    Pack of 2 - $298.86
    $298.86
    Each
       $423.99
    Pack of 2
       $298.86
    Supplier Number: 64799-02
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)