Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
About this item
All features are similar to the Potter-Elvehjem Tissue Grinder series, with additional radial serrated PTFE pestle.
This grinder has a serrated PTFE pestle that disperses homogenate into the mortar cylinder more efficiently. Depth of the serration is 0.1 to 0.15 mm (0.20 to 0.25”).
1 Options Available Below
Product Details & Documents
All features are similar to the Potter-Elvehjem Tissue Grinder series, with additional radial serrated PTFE pestle.
This grinder has a serrated PTFE pestle that disperses homogenate into the mortar cylinder more efficiently. Depth of the serration is 0.1 to 0.15 mm (0.20 to 0.25”).
Product Family Options
Product Information
- SizeAvailabilityPrice
- 102095-618Tissue Grinder PTFE with Radial SerrationsTissue Grinder PTFE with Radial Serrations2 mlEach$778.33Pack of 2$547.53Supplier Number: 64795-02
- 102095-620Tissue Grinder PTFE with Radial SerrationsTissue Grinder PTFE with Radial Serrations5 mlDiscontinued-Supplier Number: 64795-05
- 102095-622Tissue Grinder PTFE with Radial SerrationsTissue Grinder PTFE with Radial Serrations10 mlDiscontinued-Supplier Number: 64795-10
- 102095-624Tissue Grinder With Radial SerrationsTissue Grinder With Radial Serrations15 mlDiscontinued-Supplier Number: 64795-15
- 102095-626Tissue Grinder PTFE with Radial SerrationsTissue Grinder PTFE with Radial Serrations30 mlDiscontinued-Supplier Number: 64795-30
- 102095-628Tissue Grinder PTFE with Radial SerrationsTissue Grinder PTFE with Radial Serrations55 mlDiscontinued-Supplier Number: 64795-55
- 102095-630Tissue Grinder Pestle only PTFE Radial SerrationsTissue Grinder Pestle only PTFE Radial Serrations2 mlDiscontinued-Supplier Number: 64795-80
- 102095-632Tissue Grinder Pestle only PTFE Radial SerrationsTissue Grinder Pestle only PTFE Radial Serrations5 mlDiscontinued-Supplier Number: 64795-81
- 102095-634Tissue Grinder Pestle only PTFE Radial SerrationsTissue Grinder Pestle only PTFE Radial Serrations10 mlDiscontinued-Supplier Number: 64795-82
- 102095-636Tissue Grinder Pestle only PTFE Radial SerrationsTissue Grinder Pestle only PTFE Radial Serrations15 mlDiscontinued-Supplier Number: 64795-83
- 102095-638Tissue Grinder Pestle only PTFE Radial SerrationsTissue Grinder Pestle only PTFE Radial Serrations30 mlDiscontinued-Supplier Number: 64795-84
- 102095-640Tissue Grinder Pestle only PTFE Radial SerrationsTissue Grinder Pestle only PTFE Radial Serrations55 mlDiscontinued-Supplier Number: 64795-85
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)