ContactUsLinkComponent
EMS Bell Jar Shine, Electron Microscopy Sciences
EMS Bell Jar Shine, Electron Microscopy Sciences
Catalog # 102093-310
undefined
EMS Bell Jar Shine, Electron Microscopy Sciences
Catalog # 102093-310
Supplier Number:  60540-01
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Bell Jar Shine
  • Size:
    1 lb
  • Cat. no.:
    102093-310
  • Supplier No.:
    60540-01
Specifications
About this item
This unique cleaning compound will not only clean but it polishes as well. Meant to be used on any surface which is either glass, metal, porcelain, ceramic and even chrome surfaces. Perfect for the cleaning of your bell jars from carbon and sputter coaters, as well as any surfaces in the lab that are metal, ceramic, steel etc.
Delivery: Supplied in a 1 lb container.