If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Wash-N-Dry™ Slide Rack
- Cat. no.:102096-892
- Supplier No.:70366-17
Specifications
About this item
Holds 12 Slides Securely holds slides for convenient cleaning, treating, staining, and drying.
Maximum Circulation Offset design allows liquid and air to completely flow around the entire slide ensuring cleanliness and faster drying times. "Hands Off Design" Tailored to contact a minimal amount of slide surface area permitting solutions and air to affect entire slide. Fits standard slide sizes including 25 mm and 1". Laboratory Friendly Utilizes chemically and thermally resistant glass infused polypropylene. Autoclavable. Create A System Use in conjunction with a 200 mL tall form beaker to efficiently use solutions.