Orders LinkContactUsLinkComponent
EMS-Core Sampling Tools, Electron Microscopy Sciences
EMS-Core Sampling Tools, Electron Microscopy Sciences
Marketsource
EMS-Core Sampling Tools, Electron Microscopy Sciences
Marketsource
Certificates
EMS-Core Sampling Tools, Electron Microscopy Sciences
EMS-Core Sampling Tools, Electron Microscopy Sciences
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS-Core is an all-new revolutionary multi-purpose sampling tool, designed with retractable cutting cannula for easy operation.
  • Pen-like Actuator mechanically extends the cannula from the protective casing
  • Self-contained Plunger effortlessly ejects the tissue sample out of the tip
  • Polypropylene body, 304 stainless steel cutting canula (tip)
Efficient, safe, fast and precise, the EMS-Core consists of a razor-sharp stainless-steel cutting tip, designed to cut, retrieve and store cored samples from source materials such as skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates. The EMS-Core is well known and recommended as the special DNA sampling tool and is ideal for forensic and laboratory sampling purposes.
Documents
Product Family Options
Product Information
  • Availability
    Price
  • Discontinued
    Supplier Number: -
  • 103301-890
    Sampling Tool, 5 mm
    Sampling Tool, 5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.90
    $22.90
    Each
       $22.90
    Supplier Number: 69039-50
  • 103301-892
    Sampling Tool, 6 mm
    Sampling Tool, 6 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.90
    $22.90
    Each
       $22.90
    Supplier Number: 69039-60
  • 103301-894
    Sampling Tool, 7 mm
    Sampling Tool, 7 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.60
    $22.60
    Each
       $22.60
    Supplier Number: 69039-70
  • 103301-896
    Sampling Tool, 8 mm
    Sampling Tool, 8 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.82
    $22.82
    Each
       $22.82
    Supplier Number: 69039-80
  • 103302-026
    Sampling Tool, 0.35 mm
    Sampling Tool, 0.35 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.12
    $22.12
    Each
       $22.12
    Supplier Number: 69039-03
    Specifications:
    Marketsource
    Marketsource
  • 103302-296
    Sampling Tool, 0.5 mm
    Sampling Tool, 0.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.12
    $22.12
    Each
       $22.12
    Supplier Number: 69039-05
    Specifications:
    Marketsource
    Marketsource
  • 103302-298
    Sampling Tool, 0.75 mm
    Sampling Tool, 0.75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.65
    $22.65
    Each
       $22.65
    Supplier Number: 69039-07
  • 103302-300
    Sampling Tool, 1 mm
    Sampling Tool, 1 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.12
    $22.12
    Each
       $22.12
    Supplier Number: 69039-10
    Specifications:
    Marketsource
    Marketsource
  • 103302-302
    Sampling Tool, 1.5 mm
    Sampling Tool, 1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.07
    $22.07
    Each
       $22.07
    Supplier Number: 69039-15
    Specifications:
    Marketsource
    Marketsource
  • 103302-304
    Sampling Tool, 2.5 mm
    Sampling Tool, 2.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.36
    $22.36
    Each
       $22.36
    Supplier Number: 69039-25
    Specifications:
    Marketsource
    Marketsource
  • 103302-308
    Sampling Tool, 4 mm
    Sampling Tool, 4 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.36
    $22.36
    Each
       $22.36
    Supplier Number: 69039-40
    Specifications:
    Marketsource
    Marketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)