Orders LinkContactUsLinkComponent
Mini SEM Mount, Electron Microscopy Sciences
Mini SEM Mount, Electron Microscopy Sciences
Mini SEM Mount, Electron Microscopy Sciences
Certificates
Mini SEM Mount, Electron Microscopy Sciences
Mini SEM Mount, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mini Pin, Aluminum For FEI, Tescan, Zeiss, Philips, Leo, Cambridge, AMRAY, Leica, CamScan, ETEC.
Head: 6.6 x 1.3 mm, Pin: 9.5 mm.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 102098-232
    Mini Pin, Slotted Head
    Mini Pin, Slotted Head
    6.6 x 1.3 mm
    Discontinued
    Supplier Number: 75638-10
  • 102098-234
    Mini Pin, Slotted Head
    Mini Pin, Slotted Head
    6.6 x 1.3 mm
    Discontinued
    Supplier Number: 75638-20
  • 102098-236
    Mini Pin, Slotted Head
    Mini Pin, Slotted Head
    6.6 x 1.3 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $46.20
    $46.20
    Log in for availability
    Pack of 100
    Pack of 100 - $38.25
    $38.25
    Each
       $46.20
    Pack of 100
       $38.25
    Supplier Number: 75638-30
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)