Orders LinkContactUsLinkComponent
Forensic Certified Mount, Electron Microscopy Sciences
Forensic Certified Mount, Electron Microscopy Sciences
Forensic Certified Mount, Electron Microscopy Sciences
Certificates
Forensic Certified Mount, Electron Microscopy Sciences
Forensic Certified Mount, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Gun Shot residue mount is free from objectional elements that would interfere with results.
  • Made from special certified aluminum alloy
  • Mounts are 12.7 x 11 mm
  • Available with 8 or 6 mm pin
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR.
Product Family Options
Product Information
  • Pin size
    Availability
    Price
  • 102098-410
    SEM Mount
    SEM Mount
    8 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $12.31
    $12.31
    Log in for availability
    Pack of 10
    Pack of 10 - $11.22
    $11.22
    Each
       $12.31
    Pack of 10
       $11.22
    Supplier Number: 76475-05
  • 102098-412
    SEM Mount
    SEM Mount
    8 mm
    Discontinued
    Supplier Number: 76475-10
  • 102098-414
    SEM Mount
    SEM Mount
    8 mm
    Discontinued
    Supplier Number: 76475-50
  • 103301-898
    SEM Mount
    SEM Mount
    6 mm
    Log in for availability
    Log in for availability
    Each
    Each - $17.75
    $17.75
    Each
       $17.75
    Supplier Number: 76475-52
  • 103306-384
    SEM Mount
    SEM Mount
    6 mm
    Log in for availability
    Log in for availability
    Each
    Each - $507.11
    $507.11
    Each
       $507.11
    Supplier Number: 76475-54
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)